Boc-trans-4-amino-L-proline methyl ester hydrochloride salt
Lieferant: Apollo Scientific
Achtung
Synonyme: (2S,4R)-1-tert-Butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, Methyl (2S,4R)-4-aminopyrrolidine-2-carboxylate hydrochloride, N1-BOC protected
OR16416-1G OR16416-250MG OR16416-5G
APOSOR16416-1GEA 79.9 EUR
APOSOR16416-1G APOSOR16416-250MG APOSOR16416-5G
Boc-trans-4-amino-L-proline methyl ester hydrochloride salt
Boc-trans-4-amino-L-proline methyl ester hydrochloride salt
Formel: C₁₁H₂₁ClN₂O₄
Molecular Weight: 280.75 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03787929
CAS-Nummer: 334999-32-5