3-Methoxypicolinsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Methoxypyridine-2-carboxylic acid
OR16441-1G
OR16441-250MG
APOSOR16441-1GEA
104
EUR
APOSOR16441-1G
APOSOR16441-250MG
3-Methoxypicolinsäure
3-Methoxypicolinsäure
Formel:
C₇H₇NO₃ Molecular Weight: 153,1354 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07367569 CAS-Nummer: 16478-52-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...