2-Hydroxy-5-iodnicotinsäure 95%

Lieferant: Apollo Scientific

OR16444-1G OR16444-5G
APOSOR16444-1GEA 56.7 EUR
APOSOR16444-1G APOSOR16444-5G
2-Hydroxy-5-iodnicotinsäure 95%
2-Hydroxy-5-iodnicotinsäure
Formel: C₆H₅NO₃
Molecular Weight: 265,005 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11100199
CAS-Nummer: 390360-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR