2-Methylthiophene-3-carboxylic acid 98%

Lieferant: Apollo Scientific
Achtung

OR16451-1G OR16451-5G
APOSOR16451-1GEA 21.9 EUR
APOSOR16451-1G APOSOR16451-5G
2-Methylthiophene-3-carboxylic acid 98%
2-Methylthiophene-3-carboxylic acid
Formel: C₆H₆O₂S
Molecular Weight: 142,1756 g/mol
Schmelzpunkt: 115…117 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01859891
CAS-Nummer: 1918-78-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR