4-Amino-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Aminopyrazole, 1H-Pyrazol-4-amine

OR16453-10G OR16453-1G OR16453-5G
APOSOR16453-10GEA 133 EUR
APOSOR16453-10G APOSOR16453-1G APOSOR16453-5G
4-Amino-1H-pyrazole
4-Amino-1H-pyrazole
Formel: C₃H₅N₃
Molecular Weight: 83.09 g/mol
Schmelzpunkt: 72…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01693729
CAS-Nummer: 28466-26-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR