5-Amino-1-(tert-butyl)-1H-pyrrole-3-carbonitrile

Lieferant: Apollo Scientific
Achtung

OR16462-500MG
APOSOR16462-500MGEA 116 EUR
APOSOR16462-500MG
5-Amino-1-(tert-butyl)-1H-pyrrole-3-carbonitrile
5-Amino-1-(tert-butyl)-1H-pyrrole-3-carbonitrile
Formel: C₉H₁₃N₃
Schmelzpunkt: 115…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11846194
CAS-Nummer: 269726-49-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR