(3aR,6aS)-5-Benzyltetrahydropyrrolo[3,4-c]pyrrole-1,3(2H,3aH)-dione
Lieferant: Apollo Scientific
|
Achtung
|
OR16463-1G
OR16463-500MG
APOSOR16463-1GEA
538
EUR
APOSOR16463-1G
APOSOR16463-500MG
(3aR,6aS)-5-Benzyltetrahydropyrrolo[3,4-c]pyrrole-1,3(2H,3aH)-dione
(3aR,6aS)-5-Benzyltetrahydropyrrolo[3,4-c]pyrrole-1,3(2H,3aH)-dione
Formel:
C₁₃H₁₄N₂O₂ Schmelzpunkt: 157…158 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04116221 CAS-Nummer: 370879-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...