N-Methyl-1-(p-tolyl)methanamine

Lieferant: Apollo Scientific
Gefahr

Synonyme: N-Methyl-4-methylbenzylamine

OR16470-1G OR16470-25G OR16470-5G
APOSOR16470-1GEA 58 EUR
APOSOR16470-1G APOSOR16470-25G APOSOR16470-5G
N-Methyl-1-(p-tolyl)methanamine
N-Methyl-1-(p-tolyl)methanamine
Formel: C₉H₁₃N
Molecular Weight: 135,2100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115412
CAS-Nummer: 699-04-7
UN: 3265
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR