5-Cyanoindole-3-carboxyaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Formyl-1H-indole-5-carbonitrile
OR1658-1G
OR1658-5G
OR1658-10G
APOSOR1658-1GEA
21.9
EUR
APOSOR1658-1G
APOSOR1658-5G
APOSOR1658-10G
5-Cyanoindole-3-carboxyaldehyde 97%
5-Cyanoindole-3-carboxyaldehyde
Formel:
C10H6N2O Molecular Weight: 170,1674 g/mol Schmelzpunkt: 248…253 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01719101 CAS-Nummer: 17380-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...