Isoquinoline-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR16580-1G
OR16580-5G
OR16580-25G
APOSOR16580-1GEA
0
EUR
APOSOR16580-1G
APOSOR16580-5G
APOSOR16580-25G
Isoquinoline-3-carboxylic acid
Isoquinoline-3-carboxylic acid
Formel:
C₁₀H₇NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075137 CAS-Nummer: 6624-49-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...