Glycolaldehyde diethyl acetal

Lieferant: Apollo Scientific
Achtung

Synonyme: Hydroxyacetaldehyde diethyl acetal, 2,2-Diethoxyethanol

OR16597-100G OR16597-25G OR16597-5G
APOSOR16597-100GEA 299 EUR
APOSOR16597-100G APOSOR16597-25G APOSOR16597-5G
Glycolaldehyde diethyl acetal
Glycolaldehyde diethyl acetal
Formel: HOCH₂CH(OCH₂CH₃)₂
Molecular Weight: 134,18 g/mol
Siedepunkt: 166…167 °C
Flammpunkt: 62 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00051486
CAS-Nummer: 621-63-6
EINECS: 210-697-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR