1-Boc-4-bromo-3-formylindole 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Boc-4-Bromo-3-formylindole, 4-Bromo-1H-indole-3-carboxaldehyde, N-BOC protected
OR1660-1G
OR1660-5G
OR1660-10G
APOSOR1660-1GEA
29
EUR
APOSOR1660-1G
APOSOR1660-5G
APOSOR1660-10G
1-Boc-4-bromo-3-formylindole 98%
1-Boc-4-bromo-3-formylindole
Formel:
C₁₄H₁₄BrNO₃ Molecular Weight: 324,1699 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05864694 CAS-Nummer: 303041-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...