4-Amino-2-methoxypyrimidine 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methoxypyrimidin-4-amine
OR16620-1G
OR16620-5G
APOSOR16620-1GEA
29
EUR
APOSOR16620-1G
APOSOR16620-5G
4-Amino-2-methoxypyrimidine 97%
4-Amino-2-methoxypyrimidine
Formel:
C₅H₇N₃O Molecular Weight: 125,13 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00128201 CAS-Nummer: 3289-47-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...