2-Piperidino-5-pyridinboronsäurepinakolester
Lieferant: Apollo Scientific
Synonyme:
2-(Piperidin-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(Piperidin-1-yl)pyridine-5-boronic acid pinacol ester
OR16628-5G
APOSOR16628-5GEA
156
EUR
APOSOR16628-5G
2-Piperidino-5-pyridinboronsäurepinakolester
2-Piperidino-5-pyridinboronsäurepinakolester
Formel:
C₁₆H₂₅BN₂O₂ Molecular Weight: 288,1929 g/mol Schmelzpunkt: 110…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06798274 CAS-Nummer: 852228-08-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...