6-Methylindole-3-carboxyaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Methyl-1H-indole-3-carbaldehyde

OR1666-5G
APOSOR1666-5GEA 95.7 EUR
APOSOR1666-5G
6-Methylindole-3-carboxyaldehyde 95%
6-Methylindole-3-carboxyaldehyde
Formel: C₁₀H₉NO
Molecular Weight: 159,19 g/mol
Schmelzpunkt: 185…189 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00049347
CAS-Nummer: 4771-49-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR