3-Amino-1H-pyrazole-4-carboxamide 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR16676-1G
OR16676-25G
OR16676-5G
APOSOR16676-1GEA
21.9
EUR
APOSOR16676-1G
APOSOR16676-25G
APOSOR16676-5G
3-Amino-1H-pyrazole-4-carboxamide 98%
3-Amino-1H-pyrazole-4-carboxamide
Formel:
C₄H₆N₄O Molecular Weight: 126.12 g/mol Siedepunkt: 454 °C Schmelzpunkt: 187…189 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082808 CAS-Nummer: 5334-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...