(R)-(+)-Anicyphos
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4R)-(+)-5,5-Dimethyl-2-hydroxy-4-(2-methoxyphenyl)-1,3,2-dioxaphosphinane 2-oxide
OR16846-1G
OR16846-500MG
APOSOR16846-1GEA
180
EUR
APOSOR16846-1G
APOSOR16846-500MG
(R)-(+)-Anicyphos
(R)-(+)-Anicyphos
Formel:
C₁₂H₁₇O₅P Molecular Weight: 272,24 g/mol Schmelzpunkt: 197…200 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00190172 CAS-Nummer: 98674-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...