(R)-(-)-Phencyphos hydrate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4R)-(-)-5,5-Dimethyl-2-hydroxy-4-phenyl-1,3,2-dioxaphosphinane 2-oxide hydrate
OR16849-1G
OR16849-500MG
APOSOR16849-1GEA
180
EUR
APOSOR16849-1G
APOSOR16849-500MG
(R)-(-)-Phencyphos hydrate
(R)-(-)-Phencyphos hydrate
Formel:
C₁₁H₁₇O₅P Molecular Weight: 260,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150741 CAS-Nummer: 953776-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...