(R)-(+)-Chlocyphos

Lieferant: Apollo Scientific
Achtung

Synonyme: (4R)-4-(2-Chlorophenyl)-2-hydroxy-5,5-dimethyl-1,3,2-dioxaphosphinane 2-oxide, (4R)-(+)-4-(2-Chlorophenyl)-5,5-dimethyl-2-hydroxy-1,3,2-dioxaphosphinane 2-oxide

OR16850-1G OR16850-5G
APOSOR16850-1GEA 109 EUR
APOSOR16850-1G APOSOR16850-5G
(R)-(+)-Chlocyphos
(R)-(+)-Chlocyphos
Formel: C₁₁H₁₄ClO₄P
Molecular Weight: 276,66 g/mol
Schmelzpunkt: 228…234 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00069409
CAS-Nummer: 98674-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR