6-Bromo-1H-indole-2-carbaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-1H-indole-2-carboxaldehyde

OR16855-500MG
APOSOR16855-500MGEA 342 EUR
APOSOR16855-500MG
6-Bromo-1H-indole-2-carbaldehyde
6-Bromo-1H-indole-2-carbaldehyde
Formel: C₉H₆BrNO
Molecular Weight: 224,06 g/mol
Schmelzpunkt: 181…183 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06738308
CAS-Nummer: 105191-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR