3-Bromo-2-methyl-6-nitro-2H-indazole

Lieferant: Apollo Scientific

OR16857-1G OR16857-250MG
APOSOR16857-1GEA 202 EUR
APOSOR16857-1G APOSOR16857-250MG
3-Bromo-2-methyl-6-nitro-2H-indazole
3-Bromo-2-methyl-6-nitro-2H-indazole
Formel: C₈H₆BrN₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02666041
CAS-Nummer: 74209-41-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR