4-Chloro-2-(dimethylamino)nicotinaldehyde
Lieferant: Apollo Scientific
OR16861-250MG
OR16861-1G
APOSOR16861-250MGEA
24.4
EUR
APOSOR16861-250MG
APOSOR16861-1G
4-Chloro-2-(dimethylamino)nicotinaldehyde
4-Chloro-2-(dimethylamino)nicotinaldehyde
Formel:
C₈H₉ClN₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD12025925 CAS-Nummer: 1160474-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...