5-Chlor-2-formylpyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloropyridine-2-carbaldehyde, 5-Chloropicolinaldehyde, 5-Chloro-2-pyridinecarbaldehyde
OR16864-1G
OR16864-5G
OR16864-10G
APOSOR16864-1GEA
21.9
EUR
APOSOR16864-1G
APOSOR16864-5G
APOSOR16864-10G
5-Chlor-2-formylpyridin
5-Chlor-2-formylpyridin
Formel:
C₆H₄ClNO Molecular Weight: 141,56 g/mol Schmelzpunkt: 66…68 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08277271 CAS-Nummer: 31181-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...