(5-Chlor-2-pyridinyl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloro-2-(hydroxymethyl)pyridine, (5-Chloropyridin-2-yl)methanol
OR16865-1G
OR16865-250MG
OR16865-5G
APOSOR16865-1GEA
29
EUR
APOSOR16865-1G
APOSOR16865-250MG
APOSOR16865-5G
(5-Chlor-2-pyridinyl)methanol
(5-Chlor-2-pyridinyl)methanol
Formel:
C₆H₆ClNO Molecular Weight: 143,5709 g/mol Schmelzpunkt: 45…47 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD10000856 CAS-Nummer: 209526-98-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...