3-Iod-4-nitro-1H-indazol

Lieferant: Apollo Scientific
Achtung

OR16867-1G OR16867-500MG
APOSOR16867-1GEA 66.7 EUR
APOSOR16867-1G APOSOR16867-500MG
3-Iod-4-nitro-1H-indazol
3-Iodo-4-nitro-1H-indazole
Formel: C₇H₄IN₃O₂
Molecular Weight: 289,0300 g/mol
Schmelzpunkt: 210…212 °C
Lagertemperatur: Kühlschrank
MDL: MFCD07781574
CAS-Nummer: 885521-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR