Methyl-2-(4-amino-3-iodophenyl)-2-methylpropanoate
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-(4-amino-3-iodophenyl)-2-methylpropanoate
OR16873-1G
OR16873-500MG
APOSOR16873-1GEA
671
EUR
APOSOR16873-1G
APOSOR16873-500MG
Methyl-2-(4-amino-3-iodophenyl)-2-methylpropanoate
Methyl-2-(4-amino-3-iodophenyl)-2-methylpropanoate
Formel:
C₁₁H₁₄INO₂ Molecular Weight: 319,14 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12025947 CAS-Nummer: 756813-88-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...