Methyl-6-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 6-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate
OR16876-1G
OR16876-250MG
OR16876-5G
APOSOR16876-1GEA
284
EUR
APOSOR16876-1G
APOSOR16876-250MG
APOSOR16876-5G
Methyl-6-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate 95%
Methyl-6-chloro-1H-pyrrolo[2,3-b]pyridine-2-carboxylate
Formel:
C₉H₇ClN₂O₂ Molecular Weight: 210.62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12025931 CAS-Nummer: 1140512-58-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...