4-(4-Methylpiperazino)-2-(4-methylpiperidino)aniline
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(4-Methylpiperazin-1-yl)-2-(4-methylpiperidin-1-yl)aniline
OR16881-1G
OR16881-500MG
APOSOR16881-1GEA
671
EUR
APOSOR16881-1G
APOSOR16881-500MG
4-(4-Methylpiperazino)-2-(4-methylpiperidino)aniline
4-(4-Methylpiperazino)-2-(4-methylpiperidino)aniline
Formel:
C₁₇H₂₈N₄ Molecular Weight: 288,44 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12025950 CAS-Nummer: 1160474-88-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...