2,3,5-Trichlor-4-iodpyridin 95%

Lieferant: Apollo Scientific

Synonyme: 4-Iodo-2,3,5-trichloropyridine

OR16889-1G OR16889-5G
APOSOR16889-1GEA 336 EUR
APOSOR16889-1G APOSOR16889-5G
2,3,5-Trichlor-4-iodpyridin 95%
2,3,5-Trichlor-4-iodpyridin
Formel: C₅HCI₃IN
Lagertemperatur: Kühlschrank
CAS-Nummer: 406676-23-1
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR