1-(tert-Butyl) 2-ethyl 6-bromo-1H-indole-1,2-dicarboxylate

Lieferant: Apollo Scientific

Synonyme: Ethyl 6-bromo-1H-indole-2-carboxylate, N-BOC protected

OR16891-1G OR16891-500MG
APOSOR16891-1GEA 538 EUR
APOSOR16891-1G APOSOR16891-500MG
1-(tert-Butyl) 2-ethyl 6-bromo-1H-indole-1,2-dicarboxylate
1-(tert-Butyl) 2-ethyl 6-bromo-1H-indole-1,2-dicarboxylate
Formel: C₁₆H₁₈BrNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD12025933
CAS-Nummer: 939045-17-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR