2-Phenoxyisonicotinic acid 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Phenoxypyridine-4-carboxylic acid

OR16904-1G OR16904-250MG OR16904-100MG OR16904-2.5G OR16904-5G
APOSOR16904-1GEA 249 EUR
APOSOR16904-1G APOSOR16904-250MG APOSOR16904-100MG APOSOR16904-2.5G APOSOR16904-5G
2-Phenoxyisonicotinic acid 97%
2-Phenoxyisonicotinic acid
Formel: C₁₂H₉NO₃
Molecular Weight: 215,21 g/mol
Schmelzpunkt: 240…242 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09414712
CAS-Nummer: 51362-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR