5-Brom-o-kresol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Brom-2-methylphenol, 4-Brom-2-hydroxytoluol, 1-Brom-3-hydroxy-4-methylbenzol
OR16945-25G
OR16945-5G
OR16945-100G
APOSOR16945-25GEA
37.7
EUR
APOSOR16945-25G
APOSOR16945-5G
APOSOR16945-100G
5-Brom-o-kresol
5-Brom-o-kresol
Formel:
C₇H₇BrO Molecular Weight: 187,04 g/mol Schmelzpunkt: 75…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08061906 CAS-Nummer: 36138-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...