Ethyl-1H-indazol-3-carboxylat 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 1H-indazole-3-carboxylate

OR16947-5G OR16947-1G
APOSOR16947-5GEA 105 EUR
APOSOR16947-5G APOSOR16947-1G
Ethyl-1H-indazol-3-carboxylat 95+%
Ethyl-1H-indazol-3-carboxylat
Formel: C₁₀H₁₀N₂O₂
Molecular Weight: 190.20 g/mol
Schmelzpunkt: 132…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01138134
CAS-Nummer: 4498-68-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR