4-Iod-1-methyl-1H-pyrazol-5-carbonsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Iodo-1-methyl-1H-pyrazole-5-carboxylic acid

OR16973-1G OR16973-250MG OR16973-5G
APOSOR16973-1GEA 29 EUR
APOSOR16973-1G APOSOR16973-250MG APOSOR16973-5G
4-Iod-1-methyl-1H-pyrazol-5-carbonsäure 98%
4-Iod-1-methyl-1H-pyrazol-5-carbonsäure
Formel: C₅H₅IN₂O₂
Molecular Weight: 252,01 g/mol
Schmelzpunkt: 241…242 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00461121
CAS-Nummer: 75092-30-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR