5-Methyl-2-furancarbonsäure 98%
Lieferant: Apollo Scientific
Synonyme:
5-Methylfuran-2-carboxylic acid
OR16977-25G
OR16977-100G
APOSOR16977-25GEA
172
EUR
APOSOR16977-25G
APOSOR16977-100G
5-Methyl-2-furancarbonsäure 98%
5-Methyl-2-furoic acid
Formel:
C₆H₆O₃ Molecular Weight: 126,11 g/mol Schmelzpunkt: 110 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00092359 CAS-Nummer: 1917-15-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...