4(3H)-Pyrimidinone 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Pyrimidinol, 4(3H)-Pyrimidone, 3H-Pyrimidin-4-one, 4-Hydroxypyrimidine, Pyrimidin-4(3H)-one

OR16979-5G OR16979-25G OR16979-100G OR16979-500G OR16979-1KG
APOSOR16979-5GEA 21.9 EUR
APOSOR16979-5G APOSOR16979-25G APOSOR16979-100G APOSOR16979-500G APOSOR16979-1KG
4(3H)-Pyrimidinone 98%
4(3H)-Pyrimidinone
Formel: C₄H₄N₂O
Molecular Weight: 96,09 g/mol
Schmelzpunkt: 166…169 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00006664
CAS-Nummer: 4562-27-0
EINECS: 224-932-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR