2-(N-Methyl-2-pyridylamino)ethanol
Lieferant: Apollo Scientific
Synonyme:
2-(Methyl(pyridin-2-yl)amino)ethanol, N-Methyl-N-(2-pyridyl)ethanolamine, 2-[N-(2-Hydroxyethyl)-N-methylamino]pyridine
OR17029-1G
OR17029-5G
APOSOR17029-1GEA
0
EUR
APOSOR17029-1G
APOSOR17029-5G
2-(N-Methyl-2-pyridylamino)ethanol
2-(N-Methyl-2-pyridylamino)ethanol
Formel:
C₈H₁₂N₂O Molecular Weight: 152,20 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02667633 CAS-Nummer: 122321-04-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...