tert-Butyl-6-amino-1H-indazole-1-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 6-amino-1H-indazole-1-carboxylate, 6-Amino-1H-indazole, N1-BOC protected

OR17051-1G OR17051-250MG
APOSOR17051-1GEA 29 EUR
APOSOR17051-1G APOSOR17051-250MG
tert-Butyl-6-amino-1H-indazole-1-carboxylate
tert-Butyl-6-amino-1H-indazole-1-carboxylate
Formel: C₁₂H₁₅N₃O₂
Schmelzpunkt: 171…172 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04114657
CAS-Nummer: 219503-81-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR