4-Amino-5-iodopyrrolo[2,3-d]pyrimidine 95%
Lieferant: Apollo Scientific
Synonyme:
5-iodo-7H-pyrrolo[2,3-d]pyrimidin-4-amine
OR17054-1G
OR17054-5G
APOSOR17054-1GEA
78.3
EUR
APOSOR17054-1G
APOSOR17054-5G
4-Amino-5-iodopyrrolo[2,3-d]pyrimidine 95%
4-Amino-5-iodopyrrolo[2,3-d]pyrimidine
Formel:
C₆H₅IN₄ Molecular Weight: 260,04 g/mol Schmelzpunkt: 300 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD13619882 CAS-Nummer: 163622-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...