4-Amino-7H-pyrrolo[2,3-d]pyrimidine 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
7H-pyrrolo[2,3-d]pyrimidin-4-amine, 6-Amino-7-deazapurine
OR17059-5G
APOSOR17059-5GEA
31.9
EUR
APOSOR17059-5G
4-Amino-7H-pyrrolo[2,3-d]pyrimidine 98%
4-Amino-7H-pyrrolo[2,3-d]pyrimidine
Formel:
C₆H₆N₄ Molecular Weight: 134,142 g/mol Schmelzpunkt: 257…262 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01686848 CAS-Nummer: 1500-85-2 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...