4-Brom-2-pyridincarbonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-2-cyanopyridine
OR17101-100G
OR17101-5G
OR17101-25G
APOSOR17101-100GEA
928
EUR
APOSOR17101-100G
APOSOR17101-5G
APOSOR17101-25G
4-Brom-2-pyridincarbonitril 98%
4-Brom-2-pyridincarbonitril
Formel:
C₆H₃BrN₂ Schmelzpunkt: 87…91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04065805 CAS-Nummer: 62150-45-2 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...