Isochinolin-7-amin

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Aminoisoquinoline, Isoquinolin-7-amine

OR17108-1G
APOSOR17108-1GEA 63.8 EUR
APOSOR17108-1G
Isochinolin-7-amin
Isochinolin-7-amin
Formel: C₉H₈N₂
Molecular Weight: 144,1700 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08669932
CAS-Nummer: 23707-37-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR