2-Chloropyrimidine-5-boronic acid pinacol ester 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-chloro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

OR17206-5G OR17206-25G
APOSOR17206-5GEA 55.1 EUR
APOSOR17206-5G APOSOR17206-25G
2-Chloropyrimidine-5-boronic acid pinacol ester 98%
2-Chloropyrimidine-5-boronic acid pinacol ester
Formel: C₁₀H₁₄BClN₂O₂
Molecular Weight: 240,494 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11856048
CAS-Nummer: 1003845-08-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR