1-Boc-3-bromoindole 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: tert-Butyl 3-bromo-1H-indole-1-carboxylate, tert-Butyl-3-bromo-1H-indole-1-carboxylate, 3-Bromoindole-1-carboxylic acid tert-butyl ester

OR1723-1G OR1723-5G OR1723-10G
APOSOR1723-1GEA 21.9 EUR
APOSOR1723-1G APOSOR1723-5G APOSOR1723-10G
1-Boc-3-bromoindole 95%
1-Boc-3-bromoindole
Formel: C₁₃H₁₄BrNO₂
Molecular Weight: 296.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05864772
CAS-Nummer: 143259-56-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR