7-Methoxy-1-indanon

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-Methoxy-1-indanone, 7-Methoxy-2,3-dihydro-1H-inden-1-one

OR17401-1G OR17401-5G OR17401-25G
APOSOR17401-1GEA 33.5 EUR
APOSOR17401-1G APOSOR17401-5G APOSOR17401-25G
7-Methoxy-1-indanon
7-Methoxy-2,3-dihydro-1H-inden-1-one
Formel: C₁₀H₁₀O₂
Molecular Weight: 162,19 g/mol
Schmelzpunkt: 99…102 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07784339
CAS-Nummer: 34985-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR