3,5-Dichlorisonicotinsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dichloropyridine-4-carboxylic acid

OR1745-25G OR1745-5G OR1745-1G
APOSOR1745-25GEA 72.5 EUR
APOSOR1745-25G APOSOR1745-5G APOSOR1745-1G
3,5-Dichlorisonicotinsäure 98%
3,5-Dichlorisonicotinsäure
Formel: C₆H₃Cl₂NO₂
Molecular Weight: 192 g/mol
Schmelzpunkt: 230 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD01861975
CAS-Nummer: 13958-93-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR