2-Amino-3-brombenzonitril

Lieferant: Apollo Scientific
Achtung

OR17500-1G OR17500-5G OR17500-10G
APOSOR17500-1GEA 21.9 EUR
APOSOR17500-1G APOSOR17500-5G APOSOR17500-10G
2-Amino-3-brombenzonitril
2-Amino-3-brombenzonitril
Formel: C₇H₅BrN₂
Molecular Weight: 197,03 g/mol
Schmelzpunkt: 119…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD05670539
CAS-Nummer: 114344-60-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR