2-Amino-6-brombenzonitril 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR17502-5G
OR17502-100G
OR17502-25G
APOSOR17502-5GEA
24.8
EUR
APOSOR17502-5G
APOSOR17502-100G
APOSOR17502-25G
2-Amino-6-brombenzonitril 97%
2-Amino-6-brombenzonitril
Formel:
C₇H₅BrN₂ Molecular Weight: 197,02 g/mol Schmelzpunkt: 152…153 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD09834769 CAS-Nummer: 77326-62-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...