6-Brom-2-chlorchinolin

Lieferant: Apollo Scientific
Achtung

OR17558-1G OR17558-5G OR17558-10G
APOSOR17558-1GEA 21.9 EUR
APOSOR17558-1G APOSOR17558-5G APOSOR17558-10G
6-Brom-2-chlorchinolin
6-Bromo-2-chloroquinoline
Formel: C₉H₅BrClN
Molecular Weight: 242.50 g/mol
Schmelzpunkt: 148…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115272
CAS-Nummer: 1810-71-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR