1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Propylpyrazole-4-boronic acid Pinacol ester, 2-(1-Propyl-4-pyrazolyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-Propyl-1H-pyrazole-4-boronic acid, pinacol ester, 1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole

OR17570-5G
APOSOR17570-5GEA 68.3 EUR
APOSOR17570-5G
1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol
1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Formel: C₁₂H₂₁BN₂O₂
Molecular Weight: 236,1183 g/mol
Siedepunkt: 135 °C/0,065mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD05663871
CAS-Nummer: 827614-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR